Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107026.2 | NP_001100496.1 | 8, 9 | 736 | ||
XM_006246985.2 | XP_006247047.1 | 8, 9 | 782 | |||
XM_006246986.1 | XP_006247048.1 | 8, 9 | 782 | |||
GenBank mRNA | BC129123.1 | - | 8, 9 | 736 | 7130774 | |
FQ211576.1 | - | 8, 9 | 684 |