Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270838.1 | NP_001257767.1 | 3, 4 | 423 | ||
NM_001270839.1 | NP_001257768.1 | 3, 4 | 423 | |||
NM_001270840.1 | NP_001257769.1 | 3, 4 | 423 | |||
NM_022941.4 | NP_075230.1 | 3, 4 | 423 | |||
XM_006250261.2 | XP_006250323.1 | 3, 4 | 532 | |||
XM_008769758.1 | XP_008767980.1 | 3, 4 | 535 | |||
XM_008769759.1 | XP_008767981.1 | 3, 4 | 535 | |||
GenBank mRNA | AB104736.1 | - | 3, 4 | 476 | ||
AB105072.1 | - | 3, 4 | 476 | |||
AB204897.1 | - | 2, 3 | 274 | |||
AB204898.1 | - | 2, 3 | 274 | |||
AB204899.1 | - | 2, 3 | 274 | |||
AB204900.1 | - | 2, 3 | 274 | |||
AF133094.1 | - | 2, 3 | 274 | |||
AF133095.1 | - | 2, 3 | 274 | |||
BC089763.1 | - | 3, 4 | 423 | 7324318 |