Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 23, 24 | 3353 | ||
NM_001323343.1 | NP_001310272.1 | 23, 24 | 3353 | |||
NM_015446.4 | NP_056261.4 | 23, 24 | 3069 | |||
NR_136586.1 | 23, 24 | 3353 | ||||
XM_006711758.1 | XP_006711821.1 | 23, 24 | 3182 | |||
XM_011544156.2 | XP_011542458.2 | 23, 24 | 3048 | |||
XM_011544157.2 | XP_011542459.2 | 23, 24 | 3048 | |||
GenBank mRNA | AB059277.1 | - | 23, 24 | 3069 | ||
AK091510.1 | - | 1, 2 | 128 | |||
AK122844.1 | - | 7, 8 | 842 | |||
AL832213.1 | - | 13, 14 | 1481 | |||
AL833491.1 | - | 8, 9 | 953 | |||
AY157619.1 | - | 23, 24 | 3187 |
Gene Symbol: AHCTF1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077058.1 | 1 | 3168 |