Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300906.1 | NP_001287835.1 | 28 | 4355 | ||
NM_015525.3 | NP_056340.2 | 28 | 4400 | |||
XM_006715453.2 | XP_006715516.1 | 29 | 5063 | |||
XM_006715454.2 | XP_006715517.1 | 29 | 5018 | |||
GenBank mRNA | AB037838.2 | - | 27 | 4124 | ||
AF235049.1 | - | 5 | 819 | |||
AJ420588.1 | - | 3 | 284 | |||
AJ420593.1 | - | 4 | 304 | |||
AK123460.1 | - | 1 | 1120 | |||
AK126777.1 | - | 12 | 2265 | |||
AL050018.1 | - | 8 | 940 | |||
BC027490.1 | - | 6 | 584 | |||
BC038244.2 | - | 11 | 1238 | |||
BC042171.1 | - | 7 | 783 | |||
BC113696.1 | - | 27 | 3929 | 8322748 | ||
BC113698.1 | - | 27 | 3929 | 8322750 | ||
BC143736.1 | - | 26 | 3326 | 9052250 | ||
BC143745.1 | - | 27 | 3885 | 9052259 | ||
BC143748.1 | - | 25 | 3896 | |||
DQ005633.1 | - | 28 | 4400 | |||
DQ005635.1 | - | 4 | 930 |