Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130969.1 | NP_001124441.1 | 14 | 1583 | ||
NM_001130970.1 | NP_001124442.1 | 13 | 1514 | |||
NM_001130971.1 | NP_001124443.1 | 12 | 1508 | |||
NM_001178064.1 | NP_001171535.1 | 13 | 1493 | |||
NM_015537.4 | NP_056352.3 | 13 | 1577 | |||
XM_005266061.4 | XP_005266118.1 | 12 | 1510 | |||
XM_005266062.4 | XP_005266119.1 | 11 | 1441 | |||
XM_011518496.2 | XP_011516798.1 | 12 | 1447 | |||
XM_011518497.2 | XP_011516799.1 | 9 | 697 | |||
XM_017014597.1 | XP_016870086.1 | 8 | 966 | |||
GenBank mRNA | AJ420418.1 | - | 6 | 358 | ||
AK027474.1 | - | 11 | 848 | |||
AK074602.1 | - | 13 | 1545 | |||
AK292030.1 | - | 9 | 649 | |||
AK294320.1 | - | 9 | 626 | |||
AK303703.1 | - | 11 | 705 | |||
AK303976.1 | - | 9 | 744 | |||
AK316046.1 | - | 11 | 899 | |||
AL117660.2 | - | 9 | 605 | |||
AL389944.1 | - | 9 | 631 | |||
AL389945.1 | - | 7 | 1477 | |||
AL389946.1 | - | 5 | 271 | |||
AY255128.1 | - | 14 | 1583 | |||
AY255129.1 | - | 13 | 1577 | |||
AY255130.1 | - | 13 | 1514 | |||
AY255131.1 | - | 13 | 1493 | |||
AY255132.1 | - | 13 | 1819 | |||
BC004318.1 | - | 5 | 251 | |||
BC016862.2 | - | 9 | 624 | |||
BC072412.1 | - | 13 | 1562 | 6165009 | ||
BC110498.2 | - | 12 | 1375 | 40007676 |