Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN20 SIRNA Assays ›
          Gene Symbol:
          PTPN20
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 20
          Gene Aliases:
          CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
          Chromosome Location:
          Chr.10: 46911134 - 47003834 on Build GRCh38
          UniGene ID:
          Hs.440733
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s25088

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042357.4 NP_001035816.1 11 2234
          NM_001042358.4 NP_001035817.1 10 2092
          NM_001042359.3 NP_001035818.1 8 1666
          NM_001042360.3 NP_001035819.1 5 1543
          NM_001042361.4 NP_001035820.1 8 1541
          NM_001042362.3 NP_001035821.1 6 1298
          NM_001042363.4 NP_001035822.1 10 2077
          NM_001042364.4 NP_001035823.1 7 1431
          NM_001042365.3 NP_001035824.1 4 1090
          NM_001320681.1 NP_001307610.1 9 1982
          NM_001320682.1 NP_001307611.1 5 1188
          NM_001320683.1 NP_001307612.1 9 1698
          NM_001320684.1 NP_001307613.1 8 1607
          NM_001320685.1 NP_001307614.1 10 2154
          NM_001320686.1 NP_001307615.1 9 1997
          NM_001320688.1 NP_001307617.1 7 1571
          NM_001320689.1 NP_001307618.1 6 1339
          NM_001320690.1 NP_001307619.1 5 1203
          NM_001320691.1 NP_001307620.1 8 1906
          NM_015605.8 NP_056420.3 9 1683
          XM_011539605.2 XP_011537907.1 11 2378
          XM_011539606.2 XP_011537908.1 11 3470
          XM_011539607.2 XP_011537909.1 11 2369
          XM_011539610.2 XP_011537912.1 8 1852
          XM_011539611.2 XP_011537913.1 7 1852
          XM_017016042.1 XP_016871531.1 11 2315
          XM_017016044.1 XP_016871533.1 10 2135
          XM_017016045.1 XP_016871534.1 5 3454
          XM_017016046.1 XP_016871535.1 10 1997
          XM_017016049.1 XP_016871538.1 8 1728
          XM_017016050.1 XP_016871539.1 6 1595
          XM_017016053.1 XP_016871542.1 8 1571
          XM_017016054.1 XP_016871543.1 8 1584
          XM_017016055.1 XP_016871544.1 8 1673
          XM_017016056.1 XP_016871545.1 7 1579
          XM_017016057.1 XP_016871546.1 8 1889
          XM_017016058.1 XP_016871547.1 6 1615
          XM_017016059.1 XP_016871548.1 7 1573
          XM_017016060.1 XP_016871549.1 7 1419
          XM_017016061.1 XP_016871550.1 6 1369
          XM_017016062.1 XP_016871551.1 6 1489
          XM_017016063.1 XP_016871552.1 7 1460
          XM_017016064.1 XP_016871553.1 5 1212
          XM_017016065.1 XP_016871554.1 6 1262
          GenBank mRNA AL050040.1 - 9 1940
          BU567804.1 - 1 151
          BX648913.1 - 4 1091
          CR933668.1 - 5 1156

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine dephosphorylation
          protein tyrosine phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline