Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193534.1 | NP_001180463.1 | 4 | 655 | ||
NM_001193535.1 | NP_001180464.1 | 4 | 604 | |||
NM_012161.3 | NP_036293.1 | 4 | 655 | |||
NR_036464.1 | 5 | 759 | ||||
XM_006713959.3 | XP_006714022.1 | 5 | 751 | |||
XM_011513831.2 | XP_011512133.1 | 4 | 437 | |||
XM_011513832.2 | XP_011512134.1 | 4 | 584 | |||
XM_011513833.2 | XP_011512135.1 | 3 | 395 | |||
XM_017008019.1 | XP_016863508.1 | 6 | 1522 | |||
GenBank mRNA | AF142481.1 | - | 4 | 577 | ||
AF157323.1 | - | 4 | 516 | |||
AF174591.1 | - | 1 | 62 | |||
AF176700.1 | - | 4 | 539 | |||
AF199420.1 | - | 4 | 544 | |||
AK000153.1 | - | 3 | 365 | |||
AK295510.1 | - | 4 | 574 | |||
AK310860.1 | - | 4 | 584 | |||
AL832808.1 | - | 2 | 291 | |||
BC012320.1 | - | 5 | 688 | |||
BC030656.1 | - | 4 | 626 | 4837080 | ||
BI464829.1 | - | 4 | 637 | |||
BX537957.1 | - | 4 | 550 |