Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK10 SIRNA Assays ›
          Gene Symbol:
          CDK10
          Gene Name:
          cyclin dependent kinase 10
          Gene Aliases:
          PISSLRE
          Chromosome Location:
          Chr.16: 89686612 - 89696364 on Build GRCh38
          UniGene ID:
          Hs.699177
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s262

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098533.2 NP_001092003.2 7 509
          NM_001160367.1 NP_001153839.1 7 509
          NM_052987.3 NP_443713.2 7 509
          NM_052988.4 NP_443714.3 7 547
          NR_027702.1 6 474
          NR_027703.1 6 474
          XM_006721308.2 XP_006721371.1 7 603
          XM_006721310.2 XP_006721373.1 7 603
          XM_011523405.2 XP_011521707.1 7 603
          XM_011523406.2 XP_011521708.1 7 603
          XM_011523407.2 XP_011521709.1 7 603
          XM_011523408.2 XP_011521710.1 7 603
          XM_011523410.2 XP_011521712.1 7 521
          XM_011523411.2 XP_011521713.1 8 759
          XM_011523412.1 XP_011521714.1 8 729
          XM_011523413.2 XP_011521715.1 7 1161
          XM_011523414.2 XP_011521716.1 7 947
          XM_011523415.2 XP_011521717.1 6 478
          XM_011523416.2 XP_011521718.1 7 686
          XM_011523417.2 XP_011521719.1 7 603
          XM_017023806.1 XP_016879295.1 6 492
          XM_017023807.1 XP_016879296.1 7 603
          XM_017023808.1 XP_016879297.1 8 758
          XM_017023809.1 XP_016879298.1 6 478
          XM_017023810.1 XP_016879299.1 7 946
          GenBank mRNA AB209829.1 - 6 454
          AF153430.1 - 6 443
          AK131531.1 - 6 2106
          AK290485.1 - 7 467
          AK292351.1 - 7 525
          AK295397.1 - 7 513
          AK296631.1 - 6 451
          AK298386.1 - 7 547
          BC017342.2 - 7 489 4865641
          BC025301.1 - 6 442 4637860
          BC093028.1 - 7 526
          CX867290.1 - 7 512
          DB118903.1 - 6 474
          DC362478.1 - 7 547
          L33264.1 - 7 508
          X78342.1 - 7 624

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          traversing start control point of mitotic cell cycle
          negative regulation of cell proliferation
          positive regulation of MAPK cascade
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix