Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037304.1 | NP_001032381.1 | 5, 6 | 484 | ||
NM_001037305.1 | NP_001032382.1 | 5, 6 | 484 | |||
NM_014174.2 | NP_054893.1 | 4, 5 | 866 | |||
NM_199297.1 | NP_954994.1 | 5, 6 | 556 | |||
NM_199298.1 | NP_954995.1 | 5, 6 | 556 | |||
XM_005271527.3 | XP_005271584.1 | 5, 6 | 519 | |||
XM_005271528.3 | XP_005271585.1 | 5, 6 | 519 | |||
GenBank mRNA | AF059619.1 | - | 5, 6 | 528 | ||
AF087886.1 | - | 5, 6 | 489 | |||
AF161493.1 | - | 4, 5 | 828 | |||
AF182413.1 | - | 5, 6 | 495 | |||
AK308498.1 | - | 4, 5 | 745 | |||
AK311024.1 | - | 4, 5 | 848 | |||
BC006978.1 | - | 5, 6 | 540 | 3682324 | ||
BC093074.1 | - | 5, 6 | 459 | 30717693 | ||
BQ640137.1 | - | 5, 6 | 474 | |||
BU934600.1 | - | 5, 6 | 486 |