Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013358.2 | NP_037490.2 | 14 | 1711 | ||
XM_017001101.1 | XP_016856590.1 | 14 | 1614 | |||
XM_017001102.1 | XP_016856591.1 | 14 | 1614 | |||
GenBank mRNA | AB033768.2 | - | 14 | 1702 | ||
AK026652.1 | - | 14 | 1713 | |||
AK127515.1 | - | 1 | 139 | |||
AK225541.1 | - | 14 | 1713 | |||
AK293275.1 | - | 4 | 465 | |||
AK298538.1 | - | 4 | 462 | |||
AK298892.1 | - | 3 | 245 | |||
AK309106.1 | - | 1 | 76 | |||
AK314644.1 | - | 14 | 1712 | |||
BC130574.1 | - | 14 | 1690 | 40146605 | ||
BC136402.1 | - | 14 | 1690 | 9020389 |