Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 684 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 960 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 816 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1020 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1074 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 609 | |||
NM_021155.3 | NP_066978.1 | 7 | 1092 | |||
NR_026692.1 | 6 | 1192 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 709 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1034 | ||
AK293089.1 | - | 7 | 1048 | |||
AK304190.1 | - | 7 | 595 | |||
AK308595.1 | - | 4 | 635 | |||
AK313585.1 | - | 7 | 1093 | |||
AY042221.1 | - | 7 | 1025 | |||
AY042222.1 | - | 7 | 1007 | |||
AY042223.1 | - | 7 | 749 | |||
AY042224.1 | - | 6 | 317 | |||
AY042225.1 | - | 6 | 953 | |||
AY042226.1 | - | 5 | 893 | |||
AY042227.1 | - | 6 | 617 | |||
AY042229.1 | - | 6 | 1125 | |||
AY042230.1 | - | 5 | 1053 | |||
AY042231.1 | - | 5 | 915 | |||
AY042232.1 | - | 4 | 940 | |||
BC110615.2 | - | 5 | 1060 | 40032459 | ||
M98457.1 | - | 7 | 1066 |