Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDH18 SIRNA Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
CDH14, CDH14L, CDH24
Chromosome Location:
Chr.5: 19472930 - 20575902 on Build GRCh38
UniGene ID:
Hs.317632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2817
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDH18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167667.1
NP_001161139.1
8
1641
NM_001291956.1
NP_001278885.1
10
2098
NM_001291957.1
NP_001278886.1
8
1619
NM_004934.3
NP_004925.1
8
1641
XM_005248228.3
XP_005248285.1
8
1850
XM_006714435.3
XP_006714498.1
7
1457
XM_011513928.2
XP_011512230.1
6
1255
XM_011513929.2
XP_011512231.1
5
1509
XM_011513930.2
XP_011512232.1
8
1728
XM_017008924.1
XP_016864413.1
9
1969
XM_017008925.1
XP_016864414.1
9
1965
XM_017008926.1
XP_016864415.1
9
1826
XM_017008927.1
XP_016864416.1
9
1755
XM_017008928.1
XP_016864417.1
10
2013
XM_017008929.1
XP_016864418.1
7
1493
XM_017008930.1
XP_016864419.1
7
1384
XM_017008931.1
XP_016864420.1
8
1637
XM_017008932.1
XP_016864421.1
9
1688
XM_017008933.1
XP_016864422.1
9
1817
XM_017008934.1
XP_016864423.1
9
1955
XM_017008935.1
XP_016864424.1
9
1969
XM_017008936.1
XP_016864425.1
8
1850
XM_017008937.1
XP_016864426.1
8
1609
XM_017008938.1
XP_016864427.1
8
1573
XM_017008939.1
XP_016864428.1
9
1946
XM_017008940.1
XP_016864429.1
8
1834
XM_017008941.1
XP_016864430.1
8
1593
GenBank mRNA
AK289547.1
-
10
2096
AK295087.1
-
8
1582
AK308745.1
-
8
1619
BC031051.1
-
8
1597
5278717
U59325.1
-
7
1420
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0