Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDH18 SIRNA Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
CDH14, CDH14L, CDH24
Chromosome Location:
Chr.5: 19472930 - 20575902 on Build GRCh38
UniGene ID:
Hs.317632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2818
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDH18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167667.1
NP_001161139.1
13
2491
NM_001291956.1
NP_001278885.1
15
3057
NM_001291957.1
NP_001278886.1
13
2472
NM_004934.3
NP_004925.1
13
2600
XM_005248228.3
XP_005248285.1
13
2809
XM_006714435.3
XP_006714498.1
12
2416
XM_011513928.2
XP_011512230.1
11
2214
XM_011513929.2
XP_011512231.1
10
2468
XM_011513930.2
XP_011512232.1
13
2581
XM_017008924.1
XP_016864413.1
14
2928
XM_017008925.1
XP_016864414.1
14
2924
XM_017008926.1
XP_016864415.1
14
2785
XM_017008927.1
XP_016864416.1
14
2714
XM_017008928.1
XP_016864417.1
15
2972
XM_017008929.1
XP_016864418.1
12
2452
XM_017008930.1
XP_016864419.1
12
2343
XM_017008931.1
XP_016864420.1
13
2596
XM_017008932.1
XP_016864421.1
14
2647
XM_017008933.1
XP_016864422.1
14
2776
XM_017008934.1
XP_016864423.1
14
2914
XM_017008935.1
XP_016864424.1
14
2822
XM_017008936.1
XP_016864425.1
13
2703
XM_017008937.1
XP_016864426.1
13
2462
XM_017008938.1
XP_016864427.1
13
2426
XM_017008939.1
XP_016864428.1
14
2796
XM_017008940.1
XP_016864429.1
13
2684
XM_017008941.1
XP_016864430.1
13
2443
GenBank mRNA
AK289547.1
-
15
3055
BC031051.1
-
13
2556
5278717
U59325.1
-
12
2379
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0