Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282566.1 | NP_001269495.1 | 3, 4 | 392 | ||
NM_001282567.1 | NP_001269496.1 | 3, 4 | 254 | |||
NM_001282568.1 | NP_001269497.1 | 4, 5 | 513 | |||
NM_001282569.1 | NP_001269498.1 | 5, 6 | 445 | |||
NM_001282570.1 | NP_001269499.1 | 3, 4 | 254 | |||
NM_001282572.1 | NP_001269501.1 | 4, 5 | 375 | |||
NM_001282573.1 | NP_001269502.1 | 5, 6 | 445 | |||
NM_001282574.1 | NP_001269503.1 | 4, 5 | 375 | |||
NM_016505.3 | NP_057589.2 | 4, 5 | 375 | |||
XM_006710681.3 | XP_006710744.1 | 5, 6 | 519 | |||
XM_011541567.2 | XP_011539869.1 | 4, 5 | 391 | |||
GenBank mRNA | AF151077.1 | - | 4, 5 | 304 | ||
AF151085.1 | - | 4, 5 | 331 | |||
AF247661.2 | - | 3, 4 | 244 | |||
AK001929.1 | - | 4, 5 | 325 | |||
AK022506.1 | - | 4, 5 | 362 | |||
AK024049.1 | - | 4, 5 | 351 | |||
AK302248.1 | - | 5, 6 | 395 | |||
AK302343.1 | - | 3, 4 | 200 | |||
AK307478.1 | - | 3, 4 | 228 | |||
BC000685.2 | - | 4, 5 | 413 | |||
BC007446.2 | - | 4, 5 | 311 | 3009823 | ||
BC050609.1 | - | 4, 5 | 315 | 6527025 | ||
BF309574.1 | - | 4, 5 | 326 | |||
CN423139.1 | - | 4, 5 | 312 | |||
DA737244.1 | - | 5, 6 | 395 | |||
DC366453.1 | - | 4, 5 | 513 |