Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM49B SIRNA Assays ›
          Gene Symbol:
          FAM49B
          Gene Name:
          family with sequence similarity 49 member B
          Gene Aliases:
          BM-009, L1
          Chromosome Location:
          Chr.8: 129839593 - 130016697 on Build GRCh38
          UniGene ID:
          Hs.126941
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28347

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM49B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256763.1 NP_001243692.1 10 1069
          NM_016623.4 NP_057707.3 9 982
          NR_046359.1 12 1335
          NR_046360.1 12 1245
          NR_046361.1 11 1235
          XM_005250966.3 XP_005251023.1 8 838
          XM_011517108.2 XP_011515410.1 11 1131
          XM_011517109.2 XP_011515411.1 11 903
          XM_011517110.2 XP_011515412.1 10 1043
          XM_011517111.2 XP_011515413.1 10 861
          XM_011517112.2 XP_011515414.1 12 1086
          XM_011517113.2 XP_011515415.1 11 1097
          XM_011517114.2 XP_011515416.1 11 1003
          XM_011517115.2 XP_011515417.1 10 1010
          XM_011517116.2 XP_011515418.1 11 1044
          XM_011517117.2 XP_011515419.1 10 1013
          XM_011517118.2 XP_011515420.1 10 957
          XM_011517120.2 XP_011515422.1 9 956
          XM_011517121.2 XP_011515423.1 12 968
          XM_011517122.2 XP_011515424.1 10 1047
          XM_011517123.2 XP_011515425.1 10 820
          XM_011517124.2 XP_011515426.1 9 935
          XM_011517125.2 XP_011515427.1 9 874
          XM_017013539.1 XP_016869028.1 12 1090
          XM_017013540.1 XP_016869029.1 12 949
          XM_017013541.1 XP_016869030.1 10 1077
          XM_017013542.1 XP_016869031.1 12 1127
          XM_017013543.1 XP_016869032.1 12 1227
          XM_017013544.1 XP_016869033.1 12 1009
          XM_017013545.1 XP_016869034.1 13 1273
          XM_017013546.1 XP_016869035.1 11 1040
          XM_017013547.1 XP_016869036.1 13 1132
          XM_017013548.1 XP_016869037.1 11 1136
          XM_017013549.1 XP_016869038.1 11 999
          XM_017013550.1 XP_016869039.1 10 1050
          XM_017013551.1 XP_016869040.1 12 1045
          XM_017013552.1 XP_016869041.1 12 1090
          XM_017013553.1 XP_016869042.1 11 903
          XM_017013554.1 XP_016869043.1 13 1115
          XM_017013555.1 XP_016869044.1 10 1070
          XM_017013556.1 XP_016869045.1 10 816
          XM_017013557.1 XP_016869046.1 11 862
          XM_017013558.1 XP_016869047.1 11 1007
          XM_017013559.1 XP_016869048.1 11 866
          XM_017013560.1 XP_016869049.1 9 981
          XM_017013561.1 XP_016869050.1 9 733
          XM_017013562.1 XP_016869051.1 10 920
          GenBank mRNA AF208851.1 - 11 921
          AK001226.1 - 5 488
          AK002059.1 - 12 1045
          AK130372.1 - 9 869
          AY364259.1 - 6 536
          AY598320.1 - 7 668
          BC003599.1 - 10 848 3501828
          BC016345.1 - 11 989 4041187
          BC017297.2 - 12 1217 5017493
          CR749628.1 - 9 2061

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0