Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM49B SIRNA Assays ›
          Gene Symbol:
          FAM49B
          Gene Name:
          family with sequence similarity 49 member B
          Gene Aliases:
          BM-009, L1
          Chromosome Location:
          Chr.8: 129839593 - 130016697 on Build GRCh38
          UniGene ID:
          Hs.126941
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28348

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM49B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256763.1 NP_001243692.1 7 731
          NM_016623.4 NP_057707.3 6 644
          NR_046359.1 9 997
          NR_046360.1 9 907
          NR_046361.1 8 897
          XM_005250966.3 XP_005251023.1 5 500
          XM_011517108.2 XP_011515410.1 8 793
          XM_011517109.2 XP_011515411.1 8 565
          XM_011517110.2 XP_011515412.1 7 705
          XM_011517111.2 XP_011515413.1 7 523
          XM_011517112.2 XP_011515414.1 9 748
          XM_011517113.2 XP_011515415.1 8 759
          XM_011517114.2 XP_011515416.1 8 665
          XM_011517115.2 XP_011515417.1 7 672
          XM_011517116.2 XP_011515418.1 8 706
          XM_011517117.2 XP_011515419.1 7 675
          XM_011517118.2 XP_011515420.1 7 619
          XM_011517120.2 XP_011515422.1 6 618
          XM_011517121.2 XP_011515423.1 9 630
          XM_011517122.2 XP_011515424.1 7 709
          XM_011517123.2 XP_011515425.1 7 482
          XM_011517124.2 XP_011515426.1 6 597
          XM_011517125.2 XP_011515427.1 6 536
          XM_017013539.1 XP_016869028.1 9 752
          XM_017013540.1 XP_016869029.1 9 611
          XM_017013541.1 XP_016869030.1 7 739
          XM_017013542.1 XP_016869031.1 9 789
          XM_017013543.1 XP_016869032.1 9 889
          XM_017013544.1 XP_016869033.1 9 671
          XM_017013545.1 XP_016869034.1 10 935
          XM_017013546.1 XP_016869035.1 8 702
          XM_017013547.1 XP_016869036.1 10 794
          XM_017013548.1 XP_016869037.1 8 798
          XM_017013549.1 XP_016869038.1 8 661
          XM_017013550.1 XP_016869039.1 7 712
          XM_017013551.1 XP_016869040.1 9 707
          XM_017013552.1 XP_016869041.1 9 752
          XM_017013553.1 XP_016869042.1 8 565
          XM_017013554.1 XP_016869043.1 10 777
          XM_017013555.1 XP_016869044.1 7 732
          XM_017013556.1 XP_016869045.1 7 478
          XM_017013557.1 XP_016869046.1 8 524
          XM_017013558.1 XP_016869047.1 8 669
          XM_017013559.1 XP_016869048.1 8 528
          XM_017013560.1 XP_016869049.1 6 643
          XM_017013561.1 XP_016869050.1 6 395
          XM_017013562.1 XP_016869051.1 7 582
          GenBank mRNA AF208851.1 - 8 584
          AK001226.1 - 2 150
          AK002059.1 - 9 707
          AK130372.1 - 6 531
          AY364259.1 - 3 199
          AY598320.1 - 4 330
          BC003599.1 - 7 510 3501828
          BC016345.1 - 8 651 4041187
          BC017297.2 - 9 879 5017493
          BX419336.2 - 7 732
          CR749628.1 - 7 477

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc