Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28349
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
8
879
NM_016623.4
NP_057707.3
7
792
NR_046359.1
10
1145
NR_046360.1
10
1055
NR_046361.1
9
1045
XM_005250966.3
XP_005251023.1
6
648
XM_011517108.2
XP_011515410.1
9
941
XM_011517109.2
XP_011515411.1
9
713
XM_011517110.2
XP_011515412.1
8
853
XM_011517111.2
XP_011515413.1
8
671
XM_011517112.2
XP_011515414.1
10
896
XM_011517113.2
XP_011515415.1
9
907
XM_011517114.2
XP_011515416.1
9
813
XM_011517115.2
XP_011515417.1
8
820
XM_011517116.2
XP_011515418.1
9
854
XM_011517117.2
XP_011515419.1
8
823
XM_011517118.2
XP_011515420.1
8
767
XM_011517120.2
XP_011515422.1
7
766
XM_011517121.2
XP_011515423.1
10
778
XM_011517122.2
XP_011515424.1
8
857
XM_011517123.2
XP_011515425.1
8
630
XM_011517124.2
XP_011515426.1
7
745
XM_011517125.2
XP_011515427.1
7
684
XM_017013539.1
XP_016869028.1
10
900
XM_017013540.1
XP_016869029.1
10
759
XM_017013541.1
XP_016869030.1
8
887
XM_017013542.1
XP_016869031.1
10
937
XM_017013543.1
XP_016869032.1
10
1037
XM_017013544.1
XP_016869033.1
10
819
XM_017013545.1
XP_016869034.1
11
1083
XM_017013546.1
XP_016869035.1
9
850
XM_017013547.1
XP_016869036.1
11
942
XM_017013548.1
XP_016869037.1
9
946
XM_017013549.1
XP_016869038.1
9
809
XM_017013550.1
XP_016869039.1
8
860
XM_017013551.1
XP_016869040.1
10
855
XM_017013552.1
XP_016869041.1
10
900
XM_017013553.1
XP_016869042.1
9
713
XM_017013554.1
XP_016869043.1
11
925
XM_017013555.1
XP_016869044.1
8
880
XM_017013556.1
XP_016869045.1
8
626
XM_017013557.1
XP_016869046.1
9
672
XM_017013558.1
XP_016869047.1
9
817
XM_017013559.1
XP_016869048.1
9
676
XM_017013560.1
XP_016869049.1
7
791
XM_017013561.1
XP_016869050.1
7
543
XM_017013562.1
XP_016869051.1
8
730
GenBank mRNA
AF208851.1
-
9
731
AK001226.1
-
3
298
AK002059.1
-
10
855
AK130372.1
-
7
679
AY364259.1
-
4
346
AY598320.1
-
5
478
BC003599.1
-
8
658
3501828
BC016345.1
-
9
799
4041187
BC017297.2
-
10
1027
5017493
CR749628.1
-
8
625
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.1-2025.05.22-1.0