Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STYXL1 SIRNA Assays ›
Gene Symbol:
STYXL1
Gene Name:
serine/threonine/tyrosine interacting-like 1
Gene Aliases:
DUSP24, MK-STYX, MKSTYX
Chromosome Location:
Chr.7: 75995820 - 76048709 on Build GRCh38
UniGene ID:
Hs.11615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28482
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STYXL1
Interrogated Sequence
Translated Protein
RNADB ID
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317785.1
NP_001304714.1
3, 4
503
NM_001317786.1
NP_001304715.1
3, 4
353
NM_001317789.1
NP_001304718.1
2, 3
158
NM_016086.2
NP_057170.1
3, 4
500
NR_134486.1
3, 4
350
NR_134487.1
2, 3
158
XM_011516292.2
XP_011514594.1
3, 4
346
XM_011516293.2
XP_011514595.1
3, 4
346
XM_011516300.1
XP_011514602.1
3, 4
302
XM_017012296.1
XP_016867785.1
3, 4
346
XM_017012297.1
XP_016867786.1
3, 4
349
XM_017012298.1
XP_016867787.1
3, 4
309
XM_017012299.1
XP_016867788.1
3, 4
346
XM_017012300.1
XP_016867789.1
3, 4
346
XM_017012301.1
XP_016867790.1
3, 4
346
XM_017012302.1
XP_016867791.1
3, 4
349
GenBank mRNA
AF069762.1
-
3, 4
496
AF188201.1
-
1, 2
22
AF188203.1
-
1, 2
23
AF188204.1
-
1, 2
23
AK308828.1
-
3, 4
305
AK308912.1
-
3, 4
327
AL533027.3
-
3, 4
302
AY927581.1
-
LIT3580
AY927581
2, 3
139
BC024035.2
-
3, 4
463
2988900
BI772699.1
-
3, 4
353
BM126964.1
-
3, 4
234
BM824947.1
-
3, 4
474
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00211467_m1
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
intracellular signal transduction
protein tyrosine/serine/threonine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0