Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018984.3 | NP_061857.3 | 14, 15 | 1981 | ||
XM_005268984.4 | XP_005269041.1 | 14, 15 | 2692 | |||
XM_005268985.2 | XP_005269042.1 | 13, 14 | 1807 | |||
XM_011538497.1 | XP_011536799.1 | 13, 14 | 2073 | |||
XM_011538499.1 | XP_011536801.1 | 12, 13 | 1872 | |||
XM_011538500.2 | XP_011536802.1 | 13, 14 | 1897 | |||
XM_011538501.2 | XP_011536803.1 | 13, 14 | 1893 | |||
XM_017019491.1 | XP_016874980.1 | 13, 14 | 2919 | |||
XM_017019492.1 | XP_016874981.1 | 14, 15 | 2034 | |||
XM_017019493.1 | XP_016874982.1 | 12, 13 | 1778 | |||
XM_017019494.1 | XP_016874983.1 | 12, 13 | 1873 | |||
GenBank mRNA | AB037719.1 | - | 4, 5 | 1016 | ||
AB072355.1 | - | 14, 15 | 1894 | |||
BC062341.1 | - | 14, 15 | 1973 | 6376684 |