Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IP6K2 SIRNA Assays ›
          Gene Symbol:
          IP6K2
          Gene Name:
          inositol hexakisphosphate kinase 2
          Gene Aliases:
          IHPK2, PIUS
          Chromosome Location:
          Chr.3: 48688003 - 48717278 on Build GRCh38
          UniGene ID:
          Hs.595983
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s295

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IP6K2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005909.2 NP_001005909.1 2 399
          NM_001005910.2 NP_001005910.1 2 402
          NM_001005911.2 NP_001005911.1 2 399
          NM_001146178.2 NP_001139650.1 2 402
          NM_001146179.2 NP_001139651.1 2 399
          NM_001190316.1 NP_001177245.1 3 615
          NM_001190317.1 NP_001177246.1 3 538
          NM_016291.3 NP_057375.2 2 402
          NR_027437.2 2 402
          NR_027438.2 2 399
          XM_006713199.3 XP_006713262.1 3 506
          XM_006713200.1 XP_006713263.1 3 476
          XM_006713201.1 XP_006713264.1 3 553
          XM_006713202.1 XP_006713265.1 3 555
          XM_011533816.1 XP_011532118.1 3 536
          XM_011533817.1 XP_011532119.1 2 339
          XM_011533818.1 XP_011532120.1 2 342
          XM_011533822.1 XP_011532124.1 2 339
          XM_011533823.1 XP_011532125.1 2 342
          XM_017006583.1 XP_016862072.1 2 398
          XM_017006584.1 XP_016862073.1 2 373
          XM_017006585.1 XP_016862074.1 3 488
          XM_017006586.1 XP_016862075.1 3 529
          XM_017006587.1 XP_016862076.1 3 529
          XM_017006588.1 XP_016862077.1 3 555
          XM_017006589.1 XP_016862078.1 3 504
          XM_017006590.1 XP_016862079.1 3 1110
          XM_017006591.1 XP_016862080.1 3 475
          XM_017006592.1 XP_016862081.1 3 504
          XM_017006593.1 XP_016862082.1 3 504
          GenBank mRNA AF177145.1 - 1 296
          AK024045.1 - 2 339
          AK097016.1 - 8 1257
          AK290526.1 - 3 553
          AK291501.1 - 3 530
          AK304714.1 - 3 476
          AK315204.1 - 2 340
          AL137514.1 - 2 340
          AY007091.1 - 2 325
          BC001864.1 - 2 324
          BC004469.2 - 2 329
          BC019694.1 - 2 311 4941646
          BC065533.1 - 2 328 6162048
          BP262706.1 - 2 339
          BX537544.1 - 3 554
          CA488567.1 - 2 336
          CD248129.1 - 2 365
          CF139656.1 - 2 333
          CN362780.1 - 2 314
          DA083858.1 - 2 335
          DB039955.1 - 2 399

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate ion transport
          negative regulation of cell growth
          positive regulation of apoptotic process
          inositol phosphate metabolic process
          phosphatidylinositol phosphorylation
          type I interferon signaling pathway
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          protein binding
          ATP binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          inositol hexakisphosphate 1-kinase activity
          inositol hexakisphosphate 3-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0