Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321264.1 | NP_001308193.1 | 3 | 225 | ||
NM_017728.3 | NP_060198.3 | 3 | 225 | |||
XM_006721963.1 | XP_006722026.1 | 3 | 199 | |||
XM_017024797.1 | XP_016880286.1 | 3 | 168 | |||
XM_017024798.1 | XP_016880287.1 | 3 | 199 | |||
XM_017024799.1 | XP_016880288.1 | 3 | 199 | |||
GenBank mRNA | AK000262.1 | - | 3 | 208 | ||
AK074029.1 | - | 3 | 185 | |||
AK291159.1 | - | 3 | 220 | |||
BC037278.1 | - | 3 | 177 | 4825529 | ||
DN999695.1 | - | 3 | 231 |