Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30281
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
5
450
NM_001077352.1
NP_001070820.1
4
402
NM_001308044.1
NP_001294973.1
3
289
NM_018107.4
NP_060577.3
4
402
XM_011536890.2
XP_011535192.1
6
533
XM_011536892.2
XP_011535194.1
5
484
XM_011536893.2
XP_011535195.1
5
485
XM_011536894.2
XP_011535196.1
6
533
XM_011536895.2
XP_011535197.1
5
509
XM_011536896.2
XP_011535198.1
5
485
XM_011536897.2
XP_011535199.1
4
817
XM_011536900.1
XP_011535202.1
5
447
XM_011536902.1
XP_011535204.1
4
299
XM_011536903.1
XP_011535205.1
2
175
XM_011536904.1
XP_011535206.1
4
286
XM_011536905.1
XP_011535207.1
3
284
XM_011536906.1
XP_011535208.1
2
175
XM_017021398.1
XP_016876887.1
5
487
XM_017021399.1
XP_016876888.1
5
488
XM_017021400.1
XP_016876889.1
4
352
XM_017021401.1
XP_016876890.1
5
447
XM_017021402.1
XP_016876891.1
5
358
XM_017021403.1
XP_016876892.1
4
376
XM_017021404.1
XP_016876893.1
4
326
XM_017021405.1
XP_016876894.1
5
447
XM_017021406.1
XP_016876895.1
5
865
XM_017021407.1
XP_016876896.1
5
374
XM_017021408.1
XP_016876897.1
4
364
XM_017021409.1
XP_016876898.1
5
865
XM_017021410.1
XP_016876899.1
4
386
XM_017021411.1
XP_016876900.1
4
817
XM_017021412.1
XP_016876901.1
5
485
XM_017021413.1
XP_016876902.1
4
261
XM_017021414.1
XP_016876903.1
4
299
XM_017021415.1
XP_016876904.1
3
280
XM_017021416.1
XP_016876905.1
2
175
XM_017021417.1
XP_016876906.1
4
286
XM_017021418.1
XP_016876907.1
3
284
XM_017021419.1
XP_016876908.1
2
175
XM_017021420.1
XP_016876909.1
3
704
GenBank mRNA
AF087905.1
-
4
353
AF275678.1
-
5
404
AK001344.1
-
4
352
AK125593.1
-
5
412
AK225792.1
-
5
412
AK295448.1
-
4
364
AK303668.1
-
3
251
AK308055.1
-
4
365
AK315620.1
-
4
364
AL834198.1
-
5
411
BC002566.2
-
4
380
3138909
BC024208.2
-
4
339
2960467
BC106012.1
-
3
203
BX161440.1
-
5
383
CK000110.1
-
4
376
CK003276.1
-
3
281
CR457257.1
-
3
203
DC363343.1
-
4
397
KU178687.1
-
3
203
KU178688.1
-
4
251
KU178689.1
-
3
203
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline