Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304802.1 | NP_001291731.1 | 5 | 797 | ||
NM_001323894.1 | NP_001310823.1 | 5 | 711 | |||
NM_001323895.1 | NP_001310824.1 | 5 | 928 | |||
NM_001323896.1 | NP_001310825.1 | 5 | 711 | |||
NM_001323897.1 | NP_001310826.1 | 5 | 827 | |||
NM_018145.2 | NP_060615.1 | 5 | 827 | |||
XM_011521755.2 | XP_011520057.1 | 2 | 1073 | |||
GenBank mRNA | AB000782.1 | - | 4 | 566 | ||
AK001441.1 | - | 5 | 751 | |||
AK025963.1 | - | 2 | 140 | |||
AK092058.1 | - | 5 | 710 | |||
AK097286.1 | - | 6 | 639 | |||
AK123192.1 | - | 6 | 705 | |||
AK123282.1 | - | 4 | 1634 | |||
AY358793.1 | - | 4 | 567 | |||
BC008970.2 | - | 1 | 24 | |||
BC063844.1 | - | 5 | 715 | 6527521 |