Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC2A9 SIRNA Assays ›
          Gene Symbol:
          SLC2A9
          Gene Name:
          solute carrier family 2 member 9
          Gene Aliases:
          GLUT9, GLUTX, UAQTL2, URATv1
          Chromosome Location:
          Chr.4: 9771125 - 10040248 on Build GRCh38
          UniGene ID:
          Hs.444612
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s32184

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC2A9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001290.1 NP_001001290.1 8, 9 1080
          NM_020041.2 NP_064425.2 7, 8 1069
          XM_006713968.3 XP_006714031.1 7, 8 1181
          XM_006713969.2 XP_006714032.1 8, 9 1042
          XM_011513856.2 XP_011512158.1 7, 8 1206
          XM_011513857.1 XP_011512159.1 8, 9 1042
          XM_011513858.1 XP_011512160.1 8, 9 1244
          XM_011513859.2 XP_011512161.1 7, 8 1176
          XM_011513860.2 XP_011512162.1 7, 8 1176
          XM_011513861.2 XP_011512163.1 7, 8 1178
          XM_011513862.2 XP_011512164.1 6, 7 1053
          XM_011513863.2 XP_011512165.1 6, 7 2732
          XM_011513864.2 XP_011512166.1 5, 6 621
          XM_011513865.2 XP_011512167.1 7, 8 1180
          XM_011513866.2 XP_011512168.1 7, 8 1178
          XM_011513867.2 XP_011512169.1 4, 5 1135
          XM_011513868.2 XP_011512170.1 7, 8 1197
          XM_017008457.1 XP_016863946.1 7, 8 1194
          XM_017008458.1 XP_016863947.1 7, 8 1179
          XM_017008459.1 XP_016863948.1 5, 6 713
          XM_017008460.1 XP_016863949.1 6, 7 2726
          GenBank mRNA AF210317.1 - 7, 8 1062
          AK310895.1 - 8, 9 1042
          BC018897.2 - 8, 9 1080 3949549
          BC110414.1 - 7, 8 1099 5762360

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          glucose transport
          proton transport
          urate metabolic process
          organic acid transmembrane transport
          glucose transmembrane transport
          sugar:proton symporter activity
          glucose transmembrane transporter activity
          urate transmembrane transporter activity
          transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0