Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEACAM19 SIRNA Assays ›
          Gene Symbol:
          CEACAM19
          Gene Name:
          carcinoembryonic antigen related cell adhesion molecule 19
          Gene Aliases:
          CEACM19, CEAL1
          Chromosome Location:
          Chr.19: 44671452 - 44684355 on Build GRCh38
          UniGene ID:
          Hs.416925
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s32508

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEACAM19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127893.1 NP_001121365.1 2 867
          NM_020219.3 NP_064604.2 2 867
          GenBank mRNA AK027157.1 - 1 450
          AK291588.1 - 2 568
          AY358394.1 - 2 597
          BC083499.1 - 2 867 30390224
          KJ851726.1 - 2 387
          KJ851727.1 - 2 387
          KJ851728.1 - 2 387
          KJ851729.1 - 2 387
          KJ851730.1 - 2 387
          KJ851731.1 - 2 387
          KJ851732.1 - 2 387
          KJ851733.1 - 2 387
          KJ851734.1 - 2 387
          KJ851735.1 - 2 387
          KJ851736.1 - 2 387
          KJ851737.1 - 2 387
          KJ851738.1 - 2 387
          KJ851739.1 - 2 387
          KJ851740.1 - 2 387
          KJ851741.1 - 2 387
          KJ851742.1 - 2 387
          KJ851743.1 - 2 387
          KJ851744.1 - 2 387
          KJ851745.1 - 2 387
          KJ851746.1 - 2 387
          KJ851747.1 - 2 387
          KJ851748.1 - 2 387
          KJ851749.1 - 2 387
          KJ851750.1 - 2 387
          KJ851751.1 - 2 387
          KJ851752.1 - 2 387
          KJ851753.1 - 2 387
          KJ851754.1 - 2 387
          KJ851755.1 - 2 387
          KJ851756.1 - 2 387
          KJ851757.1 - 2 387
          KJ851758.1 - 2 387
          KJ851759.1 - 2 387
          KJ851760.1 - 2 387
          KJ851761.1 - 2 387
          KJ851762.1 - 2 387
          KJ851763.1 - 2 387
          KJ851764.1 - 2 387
          KJ851765.1 - 2 387
          KJ851766.1 - 2 387
          KJ851767.1 - 2 387
          KJ851768.1 - 2 387
          KJ851769.1 - 2 387
          KJ851770.1 - 2 387
          KJ851771.1 - 2 387
          KJ851772.1 - 2 387
          KJ851773.1 - 2 387
          KJ851774.1 - 2 387
          KJ851775.1 - 2 387
          KJ851776.1 - 2 387
          KJ851777.1 - 2 387
          KJ851778.1 - 2 387
          KJ851779.1 - 2 387
          KJ851780.1 - 2 387
          KJ851781.1 - 2 387
          KJ851782.1 - 2 387
          KJ851783.1 - 2 387
          KJ851784.1 - 2 387
          KJ851785.1 - 2 387
          KJ851786.1 - 2 387
          KJ851787.1 - 2 387
          KJ851788.1 - 2 387
          KJ851789.1 - 2 387
          KJ851790.1 - 2 387
          KJ851791.1 - 2 387
          KJ851792.1 - 2 387
          KJ851793.1 - 2 387
          KJ851794.1 - 2 387
          KJ851795.1 - 2 387
          KJ851796.1 - 2 387
          KJ851797.1 - 2 387

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix