Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005367.2 | NP_001005367.1 | 7 | 968 | ||
NM_001201461.1 | NP_001188390.1 | 7 | 968 | |||
NM_020659.3 | NP_065710.1 | 7 | 968 | |||
XM_017027025.1 | XP_016882514.1 | 7 | 812 | |||
XM_017027026.1 | XP_016882515.1 | 7 | 812 | |||
GenBank mRNA | AB162930.1 | - | 7 | 851 | ||
AB188496.1 | - | 7 | 968 | |||
AF177909.1 | - | 7 | 882 | |||
AK126690.1 | - | 6 | 879 | |||
AK289468.1 | - | 7 | 931 | |||
AK293468.1 | - | 9 | 1055 | |||
BC011347.1 | - | 7 | 873 | 4215136 | ||
BC019358.1 | - | 4 | 301 | 3960435 |