Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD1B SIRNA Assays ›
Gene Symbol:
GRAMD1B
Gene Name:
GRAM domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.11: 123358299 - 123627771 on Build GRCh38
UniGene ID:
Hs.144725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33113
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286563.1
NP_001273492.1
9
1199
NM_001286564.1
NP_001273493.1
8
908
NM_020716.2
NP_065767.1
8
1178
XM_005271619.4
XP_005271676.1
8
1178
XM_005271621.4
XP_005271678.1
9
5565
XM_005271623.4
XP_005271680.1
8
943
XM_005271624.4
XP_005271681.2
8
858
XM_006718892.3
XP_006718955.1
9
1095
XM_011542926.2
XP_011541228.1
9
1092
XM_011542929.2
XP_011541231.1
8
1068
XM_011542930.2
XP_011541232.1
8
1709
XM_011542931.2
XP_011541233.1
8
1623
XM_011542933.1
XP_011541235.1
6
1320
XM_017018034.1
XP_016873523.1
10
1163
XM_017018035.1
XP_016873524.1
10
1160
XM_017018036.1
XP_016873525.1
9
1630
XM_017018037.1
XP_016873526.1
10
1929
XM_017018038.1
XP_016873527.1
10
1163
XM_017018039.1
XP_016873528.1
9
1221
XM_017018040.1
XP_016873529.1
8
1017
XM_017018041.1
XP_016873530.1
9
1221
XM_017018042.1
XP_016873531.1
9
1221
XM_017018043.1
XP_016873532.1
8
854
XM_017018044.1
XP_016873533.1
8
1178
XM_017018045.1
XP_016873534.1
9
5562
XM_017018046.1
XP_016873535.1
9
1667
XM_017018047.1
XP_016873536.1
9
1664
XM_017018048.1
XP_016873537.1
8
1620
XM_017018049.1
XP_016873538.1
9
1661
XM_017018050.1
XP_016873539.1
8
937
XM_017018051.1
XP_016873540.1
6
541
GenBank mRNA
AB033027.1
-
8
841
AK127457.1
-
9
1199
AK297606.1
-
8
908
AK307795.1
-
8
1018
BC105741.1
-
8
992
40026783
BC105931.1
-
8
992
40026781
BC105932.1
-
8
992
40026782
BC107480.1
-
8
992
40026784
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0