Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267817.1 | NP_001254746.1 | 2 | 248 | ||
NM_001267818.1 | NP_001254747.1 | 2 | 248 | |||
NM_021227.3 | NP_067050.1 | 2 | 248 | |||
GenBank mRNA | AF068297.1 | - | 2 | 211 | ||
AF161425.1 | - | 2 | 232 | |||
AF201937.1 | - | 2 | 239 | |||
AF343342.1 | - | 2 | 177 | |||
AK312185.1 | - | 2 | 192 | |||
AW081061.1 | - | 2 | 249 | |||
BC016321.1 | - | 2 | 236 | 4075646 | ||
BC054857.1 | - | 2 | 238 | 4093030 | ||
BM471860.1 | - | 176 | ||||
BM541430.1 | - | 2 | 199 | |||
BM546028.1 | - | 2 | 238 | |||
BQ435510.1 | - | 248 | ||||
CB110420.1 | - | 2 | 193 | |||
CN430159.1 | - | 2 | 186 |