Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 9 | 958 | ||
XM_011544605.2 | XP_011542907.1 | 9 | 843 | |||
XM_011544606.2 | XP_011542908.1 | 9 | 894 | |||
XM_011544607.2 | XP_011542909.1 | 9 | 856 | |||
XM_017013718.1 | XP_016869207.1 | 8 | 709 | |||
XM_017013719.1 | XP_016869208.1 | 8 | 679 | |||
XM_017013720.1 | XP_016869209.1 | 8 | 794 | |||
GenBank mRNA | AK291842.1 | - | 9 | 902 | ||
AK296068.1 | - | 8 | 751 | |||
AK309020.1 | - | 2 | 75 | |||
DC332022.1 | - | 2 | 75 |