Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STRA6 SIRNA Assays ›
          Gene Symbol:
          STRA6
          Gene Name:
          stimulated by retinoic acid 6
          Gene Aliases:
          MCOPCB8, MCOPS9, PP14296
          Chromosome Location:
          Chr.15: 74179466 - 74212267 on Build GRCh38
          UniGene ID:
          Hs.24553
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s34570

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STRA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142617.1 NP_001136089.1 17, 18 1939
          NM_001142618.1 NP_001136090.1 17, 18 1921
          NM_001142619.1 NP_001136091.1 17, 18 1912
          NM_001199040.1 NP_001185969.1 17, 18 2055
          NM_001199041.1 NP_001185970.1 17, 18 2066
          NM_001199042.1 NP_001185971.1 17, 18 2170
          NM_022369.3 NP_071764.3 17, 18 1875
          XM_011521883.1 XP_011520185.1 17, 18 1841
          XM_011521884.1 XP_011520186.1 17, 18 1982
          XM_017022478.1 XP_016877967.1 17, 18 1790
          XM_017022479.1 XP_016877968.1 17, 18 1881
          XM_017022480.1 XP_016877969.1 17, 18 1905
          GenBank mRNA AF352728.1 - 17, 18 1741
          AF352729.1 - 17, 18 1851
          AF370419.2 - 17, 18 1847
          AK022603.1 - 13, 14 1425
          AK056125.1 - 17, 18 1936
          AK091152.1 - 17, 18 1965
          AK092227.1 - 12, 13 1402
          AK291966.1 - 17, 18 1811
          AK298921.1 - 17, 18 1810
          AK298928.1 - 15, 16 1479
          AK299191.1 - 17, 18 2055
          AK302932.1 - 17, 18 2066
          AK309676.1 - 17, 18 2150
          AY358748.1 - 17, 18 1851
          AY359089.1 - 17, 18 1741
          BC025256.1 - 17, 18 1847 4892821
          BX537413.1 - 17, 18 1817

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          blood vessel development
          kidney development
          pulmonary valve morphogenesis
          ventricular septum development
          heart development
          learning
          feeding behavior
          lung development
          adrenal gland development
          female genitalia development
          vocal learning
          ear development
          nose morphogenesis
          lung alveolus development
          positive regulation of behavior
          digestive tract morphogenesis
          embryonic digestive tract development
          developmental growth
          smooth muscle tissue development
          artery morphogenesis
          cognition
          neuromuscular process
          vitamin transport
          head development
          head morphogenesis
          face morphogenesis
          lung vasculature development
          diaphragm development
          embryonic camera-type eye formation
          eyelid development in camera-type eye
          uterus morphogenesis
          alveolar primary septum development
          pulmonary artery morphogenesis
          paramesonephric duct development
          ductus arteriosus closure
          receptor activity
          vitamin transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0