Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247999.1 | NP_001234928.1 | 3 | 683 | ||
NM_022349.3 | NP_071744.2 | 2 | 494 | |||
NM_152851.2 | NP_690590.1 | 2 | 494 | |||
NM_152852.2 | NP_690591.1 | 3 | 619 | |||
XM_005274177.2 | XP_005274234.1 | 3 | 678 | |||
XM_006718658.2 | XP_006718721.1 | 3 | 553 | |||
XM_006718660.2 | XP_006718723.1 | 3 | 553 | |||
XM_006718661.2 | XP_006718724.1 | 3 | 553 | |||
XM_011545209.1 | XP_011543511.1 | 3 | 553 | |||
XM_017018123.1 | XP_016873612.1 | 2 | 289 | |||
XM_017018124.1 | XP_016873613.1 | 2 | 289 | |||
XM_017018125.1 | XP_016873614.1 | 3 | 553 | |||
GenBank mRNA | AB013104.1 | - | 2 | 202 | ||
AF142409.1 | - | 2 | 222 | |||
AF172819.1 | - | 3 | 640 | |||
AF212240.1 | - | 2 | 363 | |||
AF253977.1 | - | 2 | 364 | |||
AF261136.1 | - | 2 | 361 | |||
AF350502.1 | - | 2 | 211 | |||
AF350503.1 | - | 2 | 211 | |||
AK026454.1 | - | 2 | 317 | |||
AK291870.1 | - | 3 | 619 | |||
AK300725.1 | - | 2 | 228 | |||
AK301387.1 | - | 2 | 189 | |||
BC022854.1 | - | 2 | 233 | 3934920 | ||
BC108728.1 | - | 2 | 267 | 6196213 | ||
DA917243.1 | - | 2 | 318 | |||
DA979138.1 | - | 2 | 265 |