Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTHFSD SIRNA Assays ›
          Gene Symbol:
          MTHFSD
          Gene Name:
          methenyltetrahydrofolate synthetase domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 86530176 - 86555235 on Build GRCh38
          UniGene ID:
          Hs.343627
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s34923

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTHFSD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159377.1 NP_001152849.1 2 163
          NM_001159378.1 NP_001152850.1 2 163
          NM_001159379.1 NP_001152851.1 2 160
          NM_001159380.1 NP_001152852.1 2 127
          NM_022764.2 NP_073601.2 2 160
          NR_027489.1 2 156
          NR_027490.1 2 163
          XM_005256101.1 XP_005256158.1 2 203
          XM_005256105.1 XP_005256162.1 2 209
          XM_005256106.1 XP_005256163.1 2 118
          XM_011523280.2 XP_011521582.1 2 203
          XM_011523282.2 XP_011521584.1 2 124
          XM_011523283.2 XP_011521585.1 2 124
          XM_011523284.2 XP_011521586.1 2 163
          XM_011523285.1 XP_011521587.1 2 124
          XM_011523286.1 XP_011521588.1 2 224
          XM_011523287.1 XP_011521589.1 2 128
          XM_011523289.1 XP_011521591.1 2 224
          XM_017023571.1 XP_016879060.1 2 163
          XM_017023572.1 XP_016879061.1 2 160
          XM_017023573.1 XP_016879062.1 2 117
          XM_017023574.1 XP_016879063.1 2 163
          GenBank mRNA AK023060.1 - 2 138
          AK023955.1 - 2 117
          AK096836.1 - 2 156
          AK297726.1 - 2 160
          BC013911.2 - 1 94
          BC112057.1 - 2 143 8327525
          BC112059.1 - 2 143 8327527
          BC143702.1 - 2 128 9052216
          BC143704.1 - 2 128
          BC143705.1 - 2 127
          BC143707.1 - 2 131 9052221

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0