Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD3 SIRNA Assays ›
Gene Symbol:
GRAMD3
Gene Name:
GRAM domain containing 3
Gene Aliases:
NS3TP2
Chromosome Location:
Chr.5: 126360096 - 126494377 on Build GRCh38
UniGene ID:
Hs.363558
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35303
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146319.1
NP_001139791.1
12
1368
NM_001146320.1
NP_001139792.1
11
1405
NM_001146321.1
NP_001139793.1
13
1560
NM_001146322.1
NP_001139794.1
12
1179
NM_023927.2
NP_076416.2
12
1517
XM_005272057.3
XP_005272114.1
12
1258
XM_005272058.3
XP_005272115.1
12
1255
XM_005272059.4
XP_005272116.1
13
1648
XM_005272060.3
XP_005272117.1
12
1345
XM_005272061.3
XP_005272118.1
12
1340
XM_005272062.3
XP_005272119.1
12
1306
XM_005272063.3
XP_005272120.1
12
1452
XM_005272064.3
XP_005272121.1
13
1365
XM_011543591.2
XP_011541893.1
13
2236
XM_011543592.2
XP_011541894.1
13
2229
XM_011543593.2
XP_011541895.1
12
1345
XM_011543594.2
XP_011541896.1
12
1117
XM_011543595.2
XP_011541897.1
11
1112
XM_017009781.1
XP_016865270.1
13
1123
XM_017009782.1
XP_016865271.1
13
1362
XM_017009783.1
XP_016865272.1
11
973
XM_017009784.1
XP_016865273.1
11
1164
XM_017009785.1
XP_016865274.1
11
1214
GenBank mRNA
AK024966.1
-
12
1285
AK225829.1
-
12
1285
AK293307.1
-
11
1110
AK294656.1
-
12
1170
AK296267.1
-
13
1265
AK298009.1
-
12
1102
AK298349.1
-
12
1045
AK300154.1
-
12
1313
AK303857.1
-
12
1119
AK316288.1
-
12
1332
AY116970.1
-
12
1087
BC008590.1
-
12
1250
4148772
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-Offline