Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35646
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
3, 4
1120
NM_001242533.1
NP_001229462.1
4, 5
1004
NM_001242534.1
NP_001229463.1
4, 5
866
NM_001242535.1
NP_001229464.1
4, 5
815
NM_001242536.1
NP_001229465.1
4, 5
1004
NM_001242537.1
NP_001229466.1
4, 5
815
NM_024311.3
NP_077287.1
4, 5
722
XM_011525236.2
XP_011523538.1
3, 4
384
XM_011525237.2
XP_011523539.2
4, 5
827
XM_011525238.2
XP_011523540.2
4, 5
427
XM_011525239.2
XP_011523541.2
4, 5
397
XM_011525240.2
XP_011523542.2
4, 5
423
XM_011525241.2
XP_011523543.2
4, 5
433
XM_011525242.1
XP_011523544.1
3, 4
384
XM_011525244.1
XP_011523546.1
3, 4
384
XM_017025065.1
XP_016880554.1
4, 5
455
XM_017025066.1
XP_016880555.1
4, 5
397
XM_017025067.1
XP_016880556.1
4, 5
455
XM_017025069.1
XP_016880558.1
3, 4
614
XM_017025070.1
XP_016880559.1
3, 4
614
GenBank mRNA
AF015184.1
-
4, 5
434
AF015185.1
-
4, 5
621
AF015186.1
-
4, 5
578
AK000233.1
-
3, 4
502
AK025849.1
-
3, 4
257
AK308058.1
-
4, 5
459
BC002753.2
-
4, 5
522
3629029
BC062563.1
-
4, 5
615
5140496
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_178620, BC088988, AK031384, BC137971, BC137970, and others.
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6f7b98477d-wlzpk:80/100.66.129.120:80.
git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline