Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36863
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
6
821
NM_001007471.2
NP_001007472.2
6
1169
NM_020952.4
NP_066003.3
6
821
NM_024971.5
NP_079247.5
6
821
NM_206944.3
NP_996827.3
6
821
NM_206945.3
NP_996828.3
6
821
NM_206946.3
NP_996829.3
6
821
NM_206947.3
NP_996830.3
6
821
NM_206948.2
NP_996831.1
6
821
XM_011519035.2
XP_011517337.1
6
1046
XM_011519036.2
XP_011517338.1
6
3971
XM_011519037.2
XP_011517339.1
6
3957
XM_011519038.2
XP_011517340.1
6
3954
XM_011519039.2
XP_011517341.1
6
3955
XM_011519040.2
XP_011517342.1
6
1052
XM_011519041.2
XP_011517343.1
6
3954
XM_011519042.2
XP_011517344.1
6
1052
XM_011519043.2
XP_011517345.1
6
1085
XM_011519044.2
XP_011517346.1
6
3955
XM_011519045.2
XP_011517347.1
6
1046
XM_011519046.2
XP_011517348.1
6
2749
XM_011519047.2
XP_011517349.1
6
2719
XM_017015141.1
XP_016870630.1
6
1046
XM_017015142.1
XP_016870631.1
6
1046
XM_017015143.1
XP_016870632.1
6
1046
XM_017015144.1
XP_016870633.1
6
812
XM_017015145.1
XP_016870634.1
6
1490
XM_017015146.1
XP_016870635.1
6
888
XM_017015147.1
XP_016870636.1
6
2923
XM_017015148.1
XP_016870637.1
6
1046
XM_017015149.1
XP_016870638.1
6
1046
XM_017015150.1
XP_016870639.1
6
1046
XM_017015151.1
XP_016870640.1
6
1064
XM_017015152.1
XP_016870641.1
6
2724
XM_017015153.1
XP_016870642.1
6
1056
XM_017015154.1
XP_016870643.1
6
1192
XM_017015155.1
XP_016870644.1
6
2721
XM_017015156.1
XP_016870645.1
2
244
XM_017015157.1
XP_016870646.1
6
2719
XM_017015158.1
XP_016870647.1
6
2717
XM_017015159.1
XP_016870648.1
6
812
XM_017015160.1
XP_016870649.1
6
902
XM_017015161.1
XP_016870650.1
6
1046
GenBank mRNA
AB099661.1
-
3
466
AB099662.1
-
3
466
AB099663.1
-
3
466
AB099664.1
-
3
466
AB099665.1
-
3
466
AF325212.1
-
6
792
AF536748.1
-
3
466
AF536749.1
-
3
466
AF536750.1
-
3
466
AF536751.1
-
3
466
AF536752.1
-
3
466
AF536753.1
-
3
466
AJ505025.1
-
6
1169
AJ505026.1
-
6
1169
AK021788.1
-
6
812
AK225732.1
-
6
812
AK308682.1
-
6
997
AL136545.1
-
6
1002
BC022454.2
-
5
751
BC067733.1
-
6
783
BC094699.1
-
6
815
BC121821.1
-
4
667
BC134414.1
-
4
670
BC142972.1
-
6
838
KF987075.1
-
6
877
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0