Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM107B SIRNA Assays ›
Gene Symbol:
FAM107B
Gene Name:
family with sequence similarity 107 member B
Gene Aliases:
C10orf45
Chromosome Location:
Chr.10: 14518557 - 14774897 on Build GRCh38
UniGene ID:
Hs.446315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38064
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM107B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282695.1
NP_001269624.1
4
937
NM_001282696.1
NP_001269625.1
3
413
NM_001282697.1
NP_001269626.1
2
347
NM_001282698.1
NP_001269627.1
3
661
NM_001282699.1
NP_001269628.1
2
595
NM_001282700.1
NP_001269629.1
2
313
NM_001282701.1
NP_001269630.1
2
434
NM_001282702.1
NP_001269631.1
3
500
NM_001282703.1
NP_001269632.1
2
367
NM_001320735.1
NP_001307664.1
3
379
NM_001320736.1
NP_001307665.1
3
639
NM_001320737.1
NP_001307666.1
3
433
NM_001320738.1
NP_001307667.1
3
399
NM_001320739.1
NP_001307668.1
2
399
NM_001320740.1
NP_001307669.1
2
191
NM_001320741.1
NP_001307670.1
2
292
NM_031453.3
NP_113641.2
3
871
XM_005252616.3
XP_005252673.1
3
268
XM_011519735.1
XP_011518037.1
4
385
XM_017016746.1
XP_016872235.1
3
259
XM_017016747.1
XP_016872236.1
2
236
XM_017016748.1
XP_016872237.1
2
271
XM_017016749.1
XP_016872238.1
2
333
GenBank mRNA
AK074275.1
-
1
130
AK127413.1
-
3
871
AK289959.1
-
3
661
AL136885.1
-
2
357
BC004872.2
-
1
141
BC064407.1
-
2
317
BC072452.1
-
3
413
BC144707.1
-
4
745
BF979432.1
-
2
407
CR533466.1
-
1
112
DA116139.1
-
2
191
DA269906.1
-
2
217
DA305421.1
-
3
310
DA340804.1
-
3
360
DA390740.1
-
2
354
DA409460.1
-
2
373
DA412617.1
-
3
407
DA543225.1
-
2
196
DA813465.1
-
2
220
DA922675.1
-
2
292
DA954468.1
-
2
313
DA962195.1
-
2
221
DB194372.1
-
2
333
DB198416.1
-
3
345
Back To Top
More Information
Gene Ontology Categories:
Function(s)
sensory perception of sound
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline