Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098794.1 | NP_001092264.1 | 2 | 474 | ||
NM_032127.3 | NP_115503.2 | 2 | 474 | |||
XM_006718343.2 | XP_006718406.1 | 2 | 396 | |||
XM_011520397.2 | XP_011518699.1 | 2 | 410 | |||
XM_011520398.2 | XP_011518700.1 | 2 | 455 | |||
XM_011520399.2 | XP_011518701.1 | 2 | 455 | |||
GenBank mRNA | AB051546.1 | - | 2 | 379 | ||
AK056468.1 | - | 2 | 426 | |||
AK291702.1 | - | 2 | 399 | |||
AL136723.1 | - | 2 | 408 | |||
BC030825.1 | - | 2 | 387 | 4811942 | ||
DA389621.1 | - | 2 | 474 |