Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032423.2 | NP_115799.2 | 7 | 2075 | ||
XM_005259323.1 | XP_005259380.1 | 4 | 1658 | |||
XM_006723418.2 | XP_006723481.1 | 9 | 2323 | |||
XM_011527392.1 | XP_011525694.1 | 9 | 2319 | |||
XM_011527394.2 | XP_011525696.1 | 4 | 2191 | |||
XM_017027360.1 | XP_016882849.1 | 10 | 2440 | |||
XM_017027361.1 | XP_016882850.1 | 10 | 2436 | |||
XM_017027362.1 | XP_016882851.1 | 8 | 2235 | |||
XM_017027363.1 | XP_016882852.1 | 9 | 2229 | |||
XM_017027364.1 | XP_016882853.1 | 8 | 2184 | |||
XM_017027365.1 | XP_016882854.1 | 4 | 1734 | |||
GenBank mRNA | AB058730.1 | - | 1 | 1172 | ||
AK023429.1 | - | 1 | 141 | |||
AK056552.1 | - | 8 | 2307 | |||
AL832630.2 | - | 1 | 2898 | |||
AL833325.1 | - | 7 | 2075 | |||
BC101712.1 | - | 4 | 1691 | 8069218 | ||
BC111950.1 | - | 4 | 1703 | 8327418 |