Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136494.1 | NP_001129966.1 | 4, 5 | 495 | ||
NM_001136495.1 | NP_001129967.1 | 2, 3 | 219 | |||
NM_032800.2 | NP_116189.1 | 2, 3 | 437 | |||
XM_017002599.1 | XP_016858088.1 | 3, 4 | 702 | |||
GenBank mRNA | AK027431.1 | - | 2, 3 | 412 | ||
AK096166.1 | - | 4, 5 | 495 | |||
AK292433.1 | - | 2, 3 | 219 | |||
AL713769.1 | - | 2, 3 | 186 | |||
BC066649.1 | - | 2, 3 | 425 | 6011914 | ||
DA333501.1 | - | 2, 3 | 219 | |||
DA754292.1 | - | 2, 3 | 437 |