Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009894.2 | NP_001009894.2 | 3 | 390 | ||
GenBank mRNA | AK095477.1 | - | 3 | 390 | ||
AK311153.1 | - | 2 | 230 | |||
AK311222.1 | - | 2 | 258 | |||
AL137488.1 | - | 3 | 432 | |||
BC035136.2 | - | 2 | 227 | |||
BC058014.1 | - | 2 | 255 | |||
BC092422.1 | - | 3 | 332 | 30386540 | ||
BX537868.1 | - | 3 | 362 | |||
BX640968.1 | - | 3 | 365 | |||
BX648026.1 | - | 2 | 230 | |||
CR936776.1 | - | 3 | 379 |