Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297623.2 | NP_001284552.1 | 5 | 557 | ||
NM_001322821.1 | NP_001309750.1 | 6 | 647 | |||
NM_001322823.1 | NP_001309752.1 | 6 | 589 | |||
NM_001322825.1 | NP_001309754.1 | 6 | 610 | |||
NM_001322826.1 | NP_001309755.1 | 5 | 520 | |||
NM_001322827.1 | NP_001309756.1 | 5 | 541 | |||
NM_001322828.1 | NP_001309757.1 | 6 | 655 | |||
NM_153257.4 | NP_694989.2 | 6 | 626 | |||
XM_011527486.2 | XP_011525788.1 | 2 | 269 | |||
XM_017027470.1 | XP_016882959.1 | 3 | 864 | |||
GenBank mRNA | AB209279.1 | - | 1 | 150 | ||
AK292834.1 | - | 6 | 601 | |||
AK299370.1 | - | 5 | 545 | |||
AY329493.1 | - | 6 | 610 | |||
AY500360.1 | - | 2 | 114 | |||
BC028631.1 | - | 2 | 420 | 5265256 | ||
BX649031.1 | - | 6 | 556 |