Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 3 | 505 | ||
NM_178190.2 | NP_835497.1 | 3 | 581 | |||
NM_178191.2 | NP_835498.1 | 2 | 1889 | |||
GenBank mRNA | AF114836.1 | - | 591 | |||
AK000934.1 | - | 2 | 1826 | |||
AL050386.1 | - | 2 | 1819 | |||
AL583540.3 | - | 3 | 504 | |||
BC004955.1 | - | 3 | 388 | 3625379 | ||
BC009677.1 | - | 3 | 463 | 3877506 | ||
BG036978.1 | - | 3 | 553 | |||
BG337531.1 | - | 3 | 461 | |||
BP271519.1 | - | 3 | 501 |