Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052923.1 | NP_443155.1 | 3 | 1312 | ||
XM_011514283.2 | XP_011512585.1 | 5 | 752 | |||
XM_011514284.2 | XP_011512586.1 | 5 | 708 | |||
XM_011514285.2 | XP_011512587.1 | 5 | 674 | |||
XM_011514286.2 | XP_011512588.1 | 5 | 685 | |||
XM_011514287.2 | XP_011512589.1 | 3 | 421 | |||
XM_011514288.2 | XP_011512590.1 | 2 | 209 | |||
XM_017010231.1 | XP_016865720.1 | 3 | 745 | |||
GenBank mRNA | AB067512.1 | - | 2 | 162 | ||
AK056369.1 | - | 3 | 1267 | |||
AL832486.1 | - | 3 | 427 | |||
AY517631.1 | - | 3 | 1312 | |||
BC110834.1 | - | 3 | 1040 | 6499684 | ||
BC111741.1 | - | 3 | 832 | 40038622 | ||
BC111742.1 | - | 3 | 832 | 40038617 | ||
BX647791.1 | - | 3 | 1280 |