Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF618 SIRNA Assays ›
Gene Symbol:
ZNF618
Gene Name:
zinc finger protein 618
Gene Aliases:
FP13169, NEDD10
Chromosome Location:
Chr.9: 113876282 - 114056595 on Build GRCh38
UniGene ID:
Hs.349208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41743
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF618
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318040.1
NP_001304969.1
14
2686
NM_001318041.1
NP_001304970.1
14
2689
NM_001318042.1
NP_001304971.1
15
2785
NM_133374.2
NP_588615.2
14
2506
XM_005251692.2
XP_005251749.1
17
2887
XM_005251699.2
XP_005251756.1
14
2539
XM_011518200.1
XP_011516502.1
16
2851
XM_011518201.1
XP_011516503.1
16
2827
XM_011518203.2
XP_011516505.1
16
2779
XM_011518204.2
XP_011516506.1
15
2719
XM_011518209.2
XP_011516511.1
4
1643
XM_017014241.1
XP_016869730.1
17
2884
XM_017014242.1
XP_016869731.1
16
2842
XM_017014243.1
XP_016869732.1
16
2839
XM_017014244.1
XP_016869733.1
15
2779
XM_017014245.1
XP_016869734.1
15
2734
XM_017014246.1
XP_016869735.1
15
2731
XM_017014247.1
XP_016869736.1
16
9490
XM_017014248.1
XP_016869737.1
16
2647
XM_017014249.1
XP_016869738.1
16
2644
XM_017014250.1
XP_016869739.1
15
2599
XM_017014251.1
XP_016869740.1
15
10876
XM_017014252.1
XP_016869741.1
14
2542
GenBank mRNA
AB075832.1
-
6
2699
BC012922.1
-
1
1261
BC036039.1
-
16
2788
5270564
BC053892.1
-
14
2439
BC136287.1
-
14
2634
9020274
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix