Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170710.4 | NP_733828.2 | 23 | 3345 | ||
NM_181265.3 | NP_851782.3 | 22 | 3222 | |||
XM_005262725.3 | XP_005262782.1 | 21 | 2868 | |||
XM_011531582.2 | XP_011529884.1 | 22 | 3210 | |||
XM_017007686.1 | XP_016863175.1 | 22 | 3198 | |||
XM_017007687.1 | XP_016863176.1 | 23 | 3283 | |||
XM_017007688.1 | XP_016863177.1 | 23 | 3200 | |||
XM_017007689.1 | XP_016863178.1 | 22 | 3197 | |||
XM_017007690.1 | XP_016863179.1 | 23 | 3283 | |||
XM_017007691.1 | XP_016863180.1 | 21 | 3032 | |||
XM_017007692.1 | XP_016863181.1 | 22 | 3118 | |||
XM_017007693.1 | XP_016863182.1 | 21 | 3032 | |||
XM_017007694.1 | XP_016863183.1 | 22 | 3016 | |||
XM_017007695.1 | XP_016863184.1 | 21 | 3046 | |||
XM_017007696.1 | XP_016863185.1 | 21 | 3046 | |||
GenBank mRNA | AF492460.1 | - | 23 | 3198 | ||
AK130128.1 | - | 8 | 852 | |||
BX648508.1 | - | 7 | 769 |