Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098475.1 | NP_001091945.1 | 8 | 1330 | ||
NM_182499.3 | NP_872305.3 | 8 | 1330 | |||
XM_006711156.2 | XP_006711219.1 | 3 | 960 | |||
XM_011509152.1 | XP_011507454.1 | 9 | 719 | |||
XM_011509153.1 | XP_011507455.1 | 9 | 719 | |||
XM_011509154.1 | XP_011507456.1 | 9 | 677 | |||
XM_011509155.1 | XP_011507457.1 | 9 | 719 | |||
XM_011509156.1 | XP_011507458.1 | 8 | 644 | |||
XM_011509157.1 | XP_011507459.1 | 8 | 644 | |||
XM_011509158.2 | XP_011507460.1 | 4 | 434 | |||
XM_011509159.2 | XP_011507461.1 | 4 | 425 | |||
XM_011509160.2 | XP_011507462.1 | 4 | 470 | |||
XM_011509161.1 | XP_011507463.1 | 4 | 438 | |||
XM_017000255.1 | XP_016855744.1 | 3 | 364 | |||
GenBank mRNA | AK302144.1 | - | 8 | 577 | ||
AL713777.1 | - | 3 | 236 | |||
AY358195.1 | - | 2 | 99 | |||
BC044864.1 | - | 8 | 1329 | 4836747 | ||
BC100895.1 | - | 3 | 208 | |||
BC100896.1 | - | 3 | 208 | |||
BC100897.2 | - | 3 | 208 | |||
BC100898.2 | - | 3 | 208 |