Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178457.2 | NP_848552.1 | 5 | 4381 | ||
XM_005260272.3 | XP_005260329.1 | 6 | 5320 | |||
XM_005260273.3 | XP_005260330.1 | 7 | 7394 | |||
XM_006723698.3 | XP_006723761.1 | 6 | 4717 | |||
XM_011528534.2 | XP_011526836.1 | 7 | 5961 | |||
XM_011528536.2 | XP_011526838.1 | 6 | 5143 | |||
XM_011528537.2 | XP_011526839.1 | 7 | 5404 | |||
XM_011528538.2 | XP_011526840.1 | 6 | 5486 | |||
XM_017027642.1 | XP_016883131.1 | 6 | 5775 | |||
XM_017027643.1 | XP_016883132.1 | 7 | 4801 | |||
XM_017027644.1 | XP_016883133.1 | 7 | 4839 | |||
GenBank mRNA | AL833682.1 | - | 5 | 3248 |