Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031738.2 | NP_001026908.1 | 6, 7 | 603 | ||
NR_033179.1 | 5, 6 | 535 | ||||
XM_006711930.3 | XP_006711993.1 | 6, 7 | 803 | |||
XM_006711931.3 | XP_006711994.1 | 6, 7 | 642 | |||
XM_006711932.2 | XP_006711995.1 | 4, 5 | 772 | |||
XM_011532505.2 | XP_011530807.1 | 5, 6 | 638 | |||
XM_011532507.2 | XP_011530809.1 | 5, 6 | 1427 | |||
XM_011532508.2 | XP_011530810.1 | 4, 5 | 570 | |||
XM_017003305.1 | XP_016858794.1 | 5, 6 | 1459 | |||
XM_017003306.1 | XP_016858795.1 | 6, 7 | 1192 | |||
XM_017003307.1 | XP_016858796.1 | 5, 6 | 1258 | |||
GenBank mRNA | AK074505.1 | - | 5, 6 | 536 | ||
AK098152.1 | - | 3, 4 | 1374 | |||
AK291879.1 | - | 6, 7 | 569 | |||
BC050466.1 | - | 6, 7 | 563 | 6197700 | ||
BX647409.1 | - | 5, 6 | 471 |