Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNA SIRNA Assays ›
Gene Symbol:
DTNA
Gene Name:
dystrobrevin alpha
Gene Aliases:
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location:
Chr.18: 34493290 - 34891844 on Build GRCh38
UniGene ID:
Hs.643454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4345
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128175.1
NP_001121647.1
10
1259
NM_001198938.1
NP_001185867.1
10
1259
NM_001198939.1
NP_001185868.1
10
1259
NM_001198940.1
NP_001185869.1
10
1259
NM_001198941.1
NP_001185870.1
9
1127
NM_001198942.1
NP_001185871.1
2
401
NM_001198943.1
NP_001185872.1
2
405
NM_001198944.1
NP_001185873.1
2
401
NM_001390.4
NP_001381.2
8
913
NM_001391.5
NP_001382.2
8
913
NM_001392.4
NP_001383.2
10
1263
NM_032975.3
NP_116757.2
10
1263
NM_032978.6
NP_116760.2
9
1127
NM_032979.4
NP_116761.2
10
1263
NM_032980.3
NP_116762.2
2
346
NM_032981.4
NP_116763.1
2
346
XM_005258221.4
XP_005258278.1
10
1258
XM_011525853.2
XP_011524155.1
10
1258
XM_017025575.1
XP_016881064.1
9
1127
XM_017025576.1
XP_016881065.1
10
1252
XM_017025577.1
XP_016881066.1
9
1127
XM_017025578.1
XP_016881067.1
10
1252
XM_017025579.1
XP_016881068.1
9
1127
XM_017025580.1
XP_016881069.1
10
1258
XM_017025581.1
XP_016881070.1
10
1252
XM_017025582.1
XP_016881071.1
9
1127
XM_017025583.1
XP_016881072.1
10
1258
XM_017025584.1
XP_016881073.1
10
1252
XM_017025585.1
XP_016881074.1
10
1252
XM_017025586.1
XP_016881075.1
10
1252
XM_017025587.1
XP_016881076.1
10
1252
XM_017025588.1
XP_016881077.1
9
1127
XM_017025589.1
XP_016881078.1
10
1258
XM_017025590.1
XP_016881079.1
10
1252
XM_017025591.1
XP_016881080.1
9
1127
XM_017025592.1
XP_016881081.1
10
1258
XM_017025593.1
XP_016881082.1
10
1252
XM_017025594.1
XP_016881083.1
9
1127
XM_017025595.1
XP_016881084.1
10
1258
XM_017025596.1
XP_016881085.1
10
1258
XM_017025597.1
XP_016881086.1
10
1252
XM_017025598.1
XP_016881087.1
9
1127
XM_017025599.1
XP_016881088.1
2
320
XM_017025600.1
XP_016881089.1
9
1127
XM_017025601.1
XP_016881090.1
10
1252
XM_017025602.1
XP_016881091.1
9
1127
GenBank mRNA
AB209102.1
-
10
1212
AJ009668.1
-
10
1171
AK291156.1
-
9
1062
AK295732.1
-
2
304
AK295789.1
-
2
292
AK296450.1
-
2
406
AK308410.1
-
10
1239
AK315215.1
-
2
237
AL833285.1
-
10
1219
BC005300.1
-
10
1219
3933795
BG705393.1
-
2
329
BI668892.1
-
2
323
BT006937.1
-
8
912
DC327958.1
-
2
346
DN991561.1
-
2
408
FJ535564.1
-
2
105
FJ535565.1
-
2
105
U26742.1
-
10
1236
U26743.1
-
2
293
U26744.1
-
9
970
U46744.1
-
9
976
U46745.1
-
9
1183
U46746.1
-
2
296
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001285813, NM_001285817, NM_001285808, NM_001285807, NM_001285811, and others.
Gene Ontology Categories:
Function(s)
Process(es)
striated muscle contraction
signal transduction
chemical synaptic transmission
neuromuscular synaptic transmission
protein binding
zinc ion binding
PDZ domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix