Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other XRRA1 SIRNA Assays ›
          Gene Symbol:
          XRRA1
          Gene Name:
          X-ray radiation resistance associated 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 74840910 - 74949187 on Build GRCh38
          UniGene ID:
          Hs.370145
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s44556

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: XRRA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270380.1 NP_001257309.1 11, 12 1428
          NM_001270381.1 NP_001257310.1 11, 12 1421
          NM_182969.2 NP_892014.1 14, 15 1689
          XM_005273765.2 XP_005273822.1 14, 15 1713
          XM_005273770.2 XP_005273827.1 12, 13 1493
          XM_011544755.1 XP_011543057.1 14, 15 1668
          XM_011544756.1 XP_011543058.1 13, 14 1587
          XM_011544757.1 XP_011543059.1 13, 14 1542
          XM_011544758.1 XP_011543060.1 12, 13 1542
          XM_011544759.1 XP_011543061.1 12, 13 1518
          XM_011544760.2 XP_011543062.1 14, 15 1713
          XM_011544761.1 XP_011543063.1 11, 12 1470
          XM_011544762.1 XP_011543064.1 11, 12 1452
          XM_011544763.1 XP_011543065.1 14, 15 1713
          XM_011544765.1 XP_011543067.1 12, 13 1427
          XM_011544766.2 XP_011543068.1 14, 15 1732
          XM_011544767.1 XP_011543069.1 13, 14 1634
          XM_011544768.1 XP_011543070.1 12, 13 1481
          XM_017017222.1 XP_016872711.1 13, 14 1587
          XM_017017223.1 XP_016872712.1 12, 13 1542
          XM_017017224.1 XP_016872713.1 11, 12 1452
          XM_017017226.1 XP_016872715.1 13, 14 1608
          XM_017017227.1 XP_016872716.1 13, 14 1584
          XM_017017228.1 XP_016872717.1 12, 13 1493
          XM_017017229.1 XP_016872718.1 12, 13 1437
          XM_017017230.1 XP_016872719.1 11, 12 1261
          XM_017017231.1 XP_016872720.1 10, 11 1347
          XM_017017233.1 XP_016872722.1 11, 12 1441
          XM_017017234.1 XP_016872723.1 10, 11 1326
          GenBank mRNA AF439934.1 - 3, 4 314
          AK056364.1 - 9, 10 1012
          AK074152.1 - 9, 10 1019
          BC037294.2 - 11, 12 1354
          BC040541.1 - 11, 12 1354
          BC064584.1 - 11, 12 1357 5742492
          CR749605.1 - 10, 11 1393

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to X-ray
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0